Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ PITC (Edman's Reagent) (phenylisothiocyanate)
Click to view available options
Quantity:
10 x 1 mL
Description
Thermo Scientific™ Pierce PITC (phenylisothiocyanate) is a high-purity reagent for pre-column quantitative derivatization of amino acids by reverse-phase HPLC.
Highlights:
- Also known as Edman's Reagent
- Reacts readily with amino acids in 5 to 10 minutes at room temperature
- Resulting phenylthiocarbamyl derivatives can be separated and quantified in 30 minutes using reverse-phase HPLC to produce stable products with all amino acids including proline
Ordering Alerts:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Specifications
| Product Type | PITC |
| Chemical Reactivity | Amine |
| Content And Storage | Upon receipt store at room temperature. |
| Label or Dye | Other Label or Dye |
| Quantity | 10 x 1 mL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction