Learn More
Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL)

Description
Remove fusion tags from proteins with the HRV 3C cleavage sequence with Thermo Scientific™ Pierce Human Rhinovirus (HRV) 3C Protease, a recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
Additional Information and Online Ordering
Highlights:
Effective – HRV 3C is designed to remove purification tags from your protein of interest
High activity at 4°
C – activity is ≥2000U/mg control protein; one unit will cleave 0.1mg control protein at 4°C after 16h
Flexible – dual-tagged enzyme is compatible with various columns and buffers
Includes:
Kit contains protease, cleavage buffer and control protein.
Recommended for:
Removal of purification tags from fusion proteins
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Product Line | Pierce |
| Form | Kit |
| Quantity | 1000 U |
| Product Type | Protease |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.