Learn More
Thermo Scientific™ Pierce™ 10X Western Blot Transfer Buffer, Methanol-free
Description
Thermo Scientific Pierce 10X Western Blot Transfer Buffer, Methanol-free is a space-saving stock solution for preparing the methanol-free transfer buffer used with cooled, tank (wet) Western blotting transfer appratus.
Pierce Concentrated Buffers are ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base. Simply dilute the stock solution with pure water and proceed with your experiment. The 10X Western Blot Transfer Buffer is ready to use upon dilution to 1X with water; it does not contain methanol and the addition of methanol is not necessary for use in electrophoresis.
Highlights:
- Transfer Buffer – diluted 10-fold in water, the solution is ready to use for electrophoresis (i.e., wet tank transfer from mini gels)
- Easy to use – no packets to open, no powder to dissolve, and no methanol required
- Increased accuracy – eliminates the possibility of powder remaining in a packet
- Saves time – 10X concentration eliminates time spent waiting for powder to dissolve
- Saves space – storage as concentrated stock minimizes bench space needed for solutions
Recommended for:
Wet (tank) Western blot transfer of proteins to a membrane
Specifications
Specifications
| Content And Storage | Storage: Upon receipt store at 4°C, protected from moisture. Product is shipped at ambient temperature. |
| Buffer | Transfer Buffer |
| Concentration | 10X |
| Gel Type | SDS-PAGE |
| Gel Compatibility | SDS-PAGE Gels |
| Quantity | 500 mL |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Product Type | Western Blot Transfer Buffer |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.