Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI15 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571805
Description
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].
Specifications
| PI15 | |
| Polyclonal | |
| Unconjugated | |
| PI15 | |
| peptidase inhibitor 15; CRISP8; DKFZp686F0366; P24TI; P25TI; 25 kDa trypsin inhibitor; CRISP-8; PI-15; cysteine-rich secretory protein 8; sugarCrisp | |
| Rabbit | |
| Protein A | |
| RUO | |
| 51050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.42 mg/mL | |
| PBS with 0.09% sodium azide | |
| O43692 | |
| PI15 | |
| KLH conjugated synthetic peptide between 34-63 amino acids from the N-terminal region of human PI15 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction