Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Phospholamban (Ser16, Thr17) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538317
Description
PLB is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases; whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.
Specifications
| Phospho-Phospholamban (Ser16, Thr17) | |
| Polyclonal | |
| Unconjugated | |
| PLN | |
| PLN; phospholamban; CMD1P; PLB; cardiac phospholamban; CMH18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18821, 5350, 64672 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26678, P61014, P61016 | |
| PLN | |
| A synthetic phosphopeptide derived from human Phospholamban around the phosphorylation site of Ser16 and Thr17 (R-A-SP-TP-I-E) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction