Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol (Liquid/Certified ACS), Fisher Chemical™

Catalog No. A931I500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Glass Bottle
Lined Steel Drum

Catalog No. A931I-500

Due to product restrictions, please to purchase or view availability for this product.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 42.8°C
Color Colorless
Density 1.0576g/cm³
pH 6
Boiling Point 182°C
Flash Point 79.4°C
Packaging Glass Bottle
Quantity 500 mL
UN Number 2821
Residue after Evaporation 0.05% max.
Vapor Pressure 0.35mmHg at 25°C
Percent Purity ≥89%
Grade Certified
Physical Form Liquid
Chemical Name or Material Phenol
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible liquid. Toxic by inhalation. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:4
Flammability:2
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.