Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ pH Buffer Solution
SCI_ED

Catalog No. S25849 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
pH 10.0: Blue
pH 4.0: Red
pH 7.0: Yellow
Catalog No. Quantity Packaging
S25849 500 mL pH 10.0: Blue
S25849A 500 mL pH 4.0: Red
S25849B 500 mL pH 7.0: Yellow
3 options

Catalog No. S25849

Only null left
Add to Cart
Edge
Add to Cart

Moderate Hazard

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material pH Buffer Solutions (pH 10.00)
CAS 1310-73-2, 7778-77-0, 110-44-1, 7732-18-5
Color Blue
Density 1.000
Physical Form Liquid
Quantity 500 mL
Packaging pH 10.0: Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.