Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ pH Buffer Solutions (pH 1 to pH 12), 500mL
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 mL
Packaging:
pH 1
pH 10
pH 11
pH 12
pH 2
pH 3
pH 4
pH 5
pH 6
pH 7
pH 8
pH 9
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
| Chemical Name or Material | pH Buffer Solutions (pH 1 to pH 12) |
| Color | Colorless |
| Density | ∽1g/mL |
| Quantity | 500 mL |
| Packaging | pH 6 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction