Learn More
Chemglass Life Sciences Cell Spreader, 90°, Glass
SDP
Supplier:  Chemglass Life Sciences CLS135002
Encompass_Preferred
Cell Spreader, 90¿, Glass. Made of 5mm borosilicate glass, spreader is ideal for spreading inoculums over the agar face in Petri dish cultures. Available in two styles, 90¿ bend and 120¿ bend. Sold in packages of 10 each.
Catalog No. 50-121-5126