Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Pyriftalid, MilliporeSigma™ Supelco™

Catalog No. 111012993 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mg

Catalog No. 11-101-2993

Supplier: MilliporeSigma Supelco 33694100MGR

Only null left
Add to Cart
Add to Cart

Pyriftalid is a pyrimidinyloxybenzoic herbicide used for the control of grass weeds in rice. Its mode of action involves inhibiting the biosynthesis of amino acids in weeds.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 135186-78-6
Physical Form Neat
Linear Formula C15H14N2O4 S
MDL Number MFCD06201843
Synonym 7-(4,6-Dimethoxypyrimidin-2-ylthio)-3-methylisobenzofuran-1(3 H)-one
Type Analytical Standard
Quantity 100 mg
Formula Weight 318.35
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.