Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 70% (Certified ACS), Fisher Chemical™

Catalog No. a22961lb Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 lb.
8 lb.
Packaging:
Glass Bottle
Safe-Cote™ Bottle
Safe-Cote™ Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A2296-1LB 1 lb. Safe-Cote™ Bottle
A229-8LB 8 lb. Glass Bottle
A229S-8LB 8 lb. Safe-Cote™ Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A2296-1LB
Only null left
Add to Cart

Chemical Identifiers

CAS 7601-90-3 , 7732-18-5
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.660g/cm³
Heavy Metals (as Pb) 1ppm max.
Identification Pass Test
Packaging Safe-Cote™ Bottle
Vapor Pressure 6.8mmHg at 25°C
Flash Point 113°C
Normality approx. 11.7
Assay Percent Range 69.0 to 72.0 w/w %
Quantity 1 lb.
Physical Form Liquid
UN Number 1873
Formula Weight 100.46
Percent Purity 69.0 to 72.0% (w/w)
Grade Certified ACS
Chemical Name or Material Perchloric Acid, 70%
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:3
DOTInformation DOT Class 5.1 : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.