Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ PELTOR™ WS™ LiteCom PRO III Headset
Shop All 3M Products
Click to view available options
Includes:
Hard Hat Attached with Bluetooth™
Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
Headband with Bluetooth™
Headband with Bluetooth™ and a jack to connect to external devices
Description
- Built-in programmable two-way communication radio allows for clear, hands-free communication in high levels of noise without cumbersome cables or cords.
- The headset can be programmed to use up to 70 unique two-way radio channels using analog and/or DMR digital modulation.
- Simultaneous, seamless connection to multiple external devices using Bluetooth™ technology.
- Noise attenuating ear cups help provide hearing protection in environments with potentially hazardous noise.
- Environmental listening with level dependent hearing protection for face-to-face communication and situational awareness.
- Noise-cancelling waterproof speech microphone with voice operated transmission (VOX) for hands-free operation; helps provide clear communication in noisy and demanding environments.
- User-friendly controls with voice-guided menu.
- Built tough for the working professional.
- Headset is available in headband, neckband and hard hat attached configurations.
Specifications
Specifications
| Color | Blue |
| Certifications/Compliance | Certified by CSA Group as intrinsically Safe for Use in Hazardous Locations |
| Height (Metric) | 224 mm |
| Model | MT73H7P3E4D10NA-50 |
| Weight (English) | 3 lb. |
| Weight (Metric) | 1.3 kg |
| Width (Metric) | 155 mm |
| Noise Reduction Rating (NRR) | 25 dB |
| Product Type | Protective Communication |
| Style | Hard hat attached |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction