Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Palmitoylethanolamide, 98%

Catalog No. AAJ67473MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
AAJ67473MF 50 mg
AAJ67473MA 10 mg
2 options

Catalog No. AAJ67473MF

Supplier: Thermo Scientific Chemicals J67473MF

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-31-0
Molecular Formula C18H37NO2
Synonym PEA; N-(2-Hydroxyethyl)hexadecanamide

Specifications

Quantity 50 mg
Description White
Content And Storage 2°C to 8°C
Formula Weight 299.50
Melting Point 97°C to 98°C
Product Type Palmitoylethanolamide, 98%
Form Solid
InChI Key HXYVTAGFYLMHSO-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC(=O)NCCO
IUPAC Name N-(2-hydroxyethyl)hexadecanamide
Molecular Weight (g/mol) 299.50
Assay 98%
Show More Show Less
EINECSNumber 208-867-9
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.