Learn More
BRINSEA PRODUCTS Ova-Easy Cabinet Incubators Chicken Egg Tray
Supplier: BRINSEA PRODUCTS GQ0246
This commercial style chicken egg tray is designed to provide higher egg capacity than the universal egg trays supplied with the Brinsea Ova-Easy cabinet incubators. 48 chicken or duck egg capacity per tray. Brinsea Ova-Easy Advance Series II cabinet incubators tray capacities: Ova-Easy 100 - 2 chicken egg trays; Ova-Easy 190 - 3 chicken egg trays; Ova-Easy 380 - 6 chicken egg trays; Ova-Easy 580 - 12 chicken egg trays.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.