Learn More
Bulbman Inc Osram BRL Lamp, Clear, Filament, 50 Watt, 12 Volt
SDP
Supplier:  Bulbman Inc 102308
Encompass_Preferred
Lamp, Osram BRL, Clear, Filament: CBar6, Avg Life: 50, Lumens: 1600, Bulb Type: T3.5, G6.35 Base, 50w, 12V, Max Overall Length: 1.720 in., Light Center Length: 1.170 in., CRI: 100, LIF: A1/220, Orientation: VBD
Catalog No. NC9088220