Learn More
Selleck Chemical LLC Iadademstat (ORY-1001) 2HCl 5mg 1431326-61-2 RG-6016
Supplier: Selleck Chemical LLC S77955MG
ORY-1001 (RG-6016) 2HCl is an orally active and selective lysine-specific demethylase LSD1/KDM1A inhibitor with IC50 of <20 nM, with high selectivity against related FAD dependent aminoxidases. Phase 1. *For Research & Development use only. Product is not intended for drug, household, or human consumption.