Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR4L1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | OR4L1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OR4L1 Polyclonal specifically detects OR4L1 in Human samples. It is validated for Western Blot.Specifications
| OR4L1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| olfactory receptor 4L1, Olfactory receptor 4L2, Olfactory receptor OR14-28, olfactory receptor, family 4, subfamily L, member 1, olfactory receptor, family 4, subfamily L, member 2 pseudogene, OR14-28, OR4L2P | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR4L1 (NP_001004717). Peptide sequence CIFIYVWPFSSLASNKTLAVFYTVITPLLNPSIYTLRNKKMQEAIRKLRF | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 122742 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title