Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Open Head Steel Drum
SDP

Catalog No. 18929B
Change view
Click to view available options
Capacity (Metric):
18.92 L
114 L
208.19 L
321.76 L
Color:
Black
Black, White
Gray
Yellow
Material:
Carbon Steel
Steel
Certifications/Compliance:
UN 1A2/X428/S-DOT/UN
UN 1A2/Y1.6/150/X400/S-DOT/UN
UN 1A2/Y27/S-DOT/UN
UN A2/Y1.5/150, UN 1A2/X400/S-DOT/UN
UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN
Catalog No. Capacity (Metric) Color Material Certifications/Compliance
18-929B 208.19 L Black Carbon Steel UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN
18-931A 18.92 L Gray Steel UN 1A2/Y27/S-DOT/UN
19-130-4692 208.19 L Black, White Steel UN 1A2/Y1.6/150/X400/S-DOT/UN
18-930C 114 L Black Carbon Steel UN A2/Y1.5/150, UN 1A2/X400/S-DOT/UN
19-130-4693 321.76 L Yellow Steel UN 1A2/X428/S-DOT/UN
19-130-4849 208.19 L Black Steel UN 1A2/Y1.6/150/X400/S-DOT/UN
6 options

Catalog No. 18-929B

Supplier: Youngstown Barrel & Drum 194

Only null left
Add to Cart
Add to Cart

Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.

Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.

With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.

  • Salvage drum designed to overpack and ship damaged and/or leaking drums
  • Also qualify for direct containment of hazardous solids
  • Freight Class 50

Specifications

Certifications/Compliance UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN
Product Type Unlined Drum
Closure Type Bolt Ring
Description 18 Gauge
Capacity (Metric) 208.19 L
Color Black
For Use With (Application) For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums
Material Carbon Steel
Freight Class 50
Head Style Open
Quantity Ea.
Height (English) 33.25 in.
Height (Metric) 84.45 cm
Diameter (English) Inner 22.5 in.
Diameter (Metric) Inner 57.15 cm
Weight (English) 49 lb.
Weight (Metric) 22.22 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.