Learn More
Sigma Aldrich Fine Chemicals Biosciences OhausR ExplorerR semi micr1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z7439241EA
            ApplicationsWeighing Percent Weighing Parts Counting Check Weighing Dynamic/Animal Weighing Filling Totalization Formulation Differential Weighing Density Determination Peak Hold Pipette Adjustment SQCDisplayFull-color VGA graphic display (5.7 in. diagonal) 4-wire resistive touch screen. QWERTY keyboard and numeric keypad to quickly input GLP and GMP data and other application dataOperationAC Adapter (Included)CommunicationEasy access communication ports including 2 units of USB port and RS232 and an optional fourth port of either RS232 or Ethernet. GLP/GMP Data Ouput with Real Time Clock. Data Transfer Function.ConstructionMetal base ABS top housing stainless steel pananti-static glass draftshield with flip-top door replaceable in-use cover automatic door as option modelsDesign FeaturesFully-automatic internal calibration system fast stabilization time four touchless sensors built-in ionizer with automatic door models up to 14 operating languages OIML models ava