Learn More
Euthanex Oxygen Regulator with E-Cylinder Fitting
Supplier: Euthanex EZ330
Oxygen Regulator with E-Cylinder Fitting. Factory preset to deliver 50 PSI directly from portable oxygen tanks ('E' Cylinders) using a CGA-870 yoke connection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.