Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Isobutylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. I03871G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. I03871G

Supplier: TCI America I03871G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6084-58-8
Molecular Formula C4H12ClNO
Molecular Weight (g/mol) 125.596
MDL Number MFCD00060208
InChI Key WOXGREIMSJFDPG-UHFFFAOYSA-N
Synonym Isobutyloxyamine Hydrochloride
PubChem CID 12696238
IUPAC Name O-(2-methylpropyl)hydroxylamine;hydrochloride
SMILES CC(C)CON.Cl

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
Formula Weight 125.60
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material O-Isobutylhydroxylamine Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.