Learn More
Research Products International Corp Nystatin, 10 Grams
SDP
Supplier:  Research Products International Corp N8202010.0
Encompass_Preferred
Nystatin is an antifungal polyene antibiotic that is effective against a wide range of fungi, including yeast and molds. It works by binding to the fungal cell membrane, disrupting its integrity and leading to leakage of cellular contents. 
Nystatin is particularly useful in preventing and treating fungal contamination in cell cultures, making it a valuable tool in microbiological research. Researchers often use Nystatin to maintain aseptic conditions in cell culture experiments.
It is added to growth media to prevent the overgrowth of fungi that could potentially compromise the integrity of cell lines and experimental results. It is selective for fungi and has minimal toxicity to mammalian cells at effective antifungal concentrations.
Catalog No. 50-213-583
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.