Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Nucleic Acid Testing for COVID-19 with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672274 Shop All EDVOTEK Products
Click to view available options
Class Size:
8 Lab Groups

Catalog No. 01-672-274

Supplier: EDVOTEK™ 123S

Only null left
Add to Cart
Add to Cart

In this simulated Nucleic Acid Testing for COVID-19 with SYBR™ Safe kit, we will use electrophoresis to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Hands-on simulation of molecular testing for COVID-19
  • Students learn about the characteristics of coronaviruses
  • Electrophoresis is used to analyze simulated RT-PCR samples
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.