Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Pore Complex Monoclonal Antibody (39C7)
GREENER_CHOICE

Catalog No. PIMA533097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533097 Supplier Invitrogen™ Supplier No. MA533097
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Nuclear Pore Complex Monoclonal Antibody for ICC/IF

The nuclear pore complex (NPC) serves as the sole bidirectional gateway of macromolecules in and out of the nucleus. Each NPC consists of ~1,000 protein subunits, termed nucleoporins, and totals to a molecular mass of ~110 Mda. NPCs maintain the integrity of the nuclear compartment by preventing macromolecules from freely diffusing in or out of the nucleus. (Annu Rev Biochem, 2019. PMID 30883195).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Pore Complex
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 39C7
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation tissue culture supernatant with 10mM sodium azide
Gene NUP155
Gene Accession No. O08587, O08658, O75694, P11654, P17955, P35658, P37198, P37199, P49790, P49791, P49793, P52590, P52948, P57740, Q12769, Q4QQS8, Q63850, Q66HC5, Q68FY1, Q6PFD9, Q80U93, Q8BH74, Q8BJ71, Q8CEC0, Q8N1F7, Q8NFH5, Q8R0G9, Q8R480, Q8R4R6, Q8TEM1, Q8WUM0, Q92621, Q99567, Q99P88, Q9BW27, Q9JIH2, Q9QY81, Q9UKX7, Q9Z0W3
Gene Alias 107 kDa nucleoporin; 133 kDa nucleoporin; 153 kDa nucleoporin; 155 kDa nucleoporin; 160 kDa nucleoporin; 160kDa; 1700030K07Rik; 205 kDa nucleoporin; 214 kDa nucleoporin; 2310006I24Rik; 2410008G02Rik; 2810011M03Rik; 35 kDa nucleoporin; 35kDa; 3830404O05Rik; 4732457F17; 50 kDa nucleoporin; 5330402E05Rik; 62 kDa nucleoporin; 85 kDa nucleoporin; 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; 93 kDa nucleoporin; 96 kDa nucleoporin; 98 kDa nucleoporin; 9830001L10; AA414952; AA589433; ADAR2; ADIR2; AI413123; AI426861; AI790512; AI836801; AI849286; ATFB15; AU020188; AU045898; AV248391; AW541137; B130015D15Rik; BC039282; C76801; C7orf14; C88147; CAIN; CAN; CAN protein, putative oncogene; CBP-interacting protein 4; Cip4; D2H9S46E; D7Ertd649e; D930027M19Rik; FROUNT; gene trap locus 1-13 protein; gene trap locus-13; GLFG-repeat containing nucleoporin; gp190; GP210; Gtl1-13; Gtl13; Gtl-13; hNUP133; HNUP153; IBSN; karyoporin; KIAA0023; Kiaa0095; Kiaa0197; KIAA0225; KIAA0791; KIAA0906; mermaid; Mitotic phosphoprotein 44; mKIAA0197; mKIAA0225; mKIAA0791; Mp44; MP-44; N153; N155; NIC96; NO44; NP44; Np62; NPAP60; NPAP60L; NPHS11; NPHS12; NPHS13; Nuclear envelope pore membrane protein POM 210; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex 1; nuclear pore complex protein 88; nuclear pore complex protein hnup153; nuclear pore complex protein Nup107; Nuclear pore complex protein Nup133; nuclear pore complex protein Nup153; Nuclear pore complex protein Nup155; nuclear pore complex protein Nup160; Nuclear pore complex protein Nup205; nuclear pore complex protein Nup214; Nuclear pore complex protein Nup50; nuclear pore complex protein Nup53; Nuclear pore complex protein Nup85; nuclear pore complex protein Nup88; Nuclear pore complex protein Nup93; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; nuclear pore complex protein Nup98; nuclear pore complex protein Nup98-Nup96; nuclear pore complex protein Nup98-Nup96; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; Nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 107; nucleoporin 107kDa; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin 153; nucleoporin 153kD; nucleoporin 153kDa; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin 160; nucleoporin 160kD; nucleoporin 160kDa; nucleoporin 205; nucleoporin 205kDa; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 214; nucleoporin 214kDa; nucleoporin 35; nucleoporin 35kDa; nucleoporin 50; nucleoporin 50 kDa; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin 85; nucleoporin 85kDa; nucleoporin 88; nucleoporin 88kDa; nucleoporin 93; nucleoporin 93kDa; nucleoporin 98; nucleoporin 98kD; nucleoporin 98kDa; nucleoporin Nup107; Nucleoporin Nup133; nucleoporin Nup153; Nucleoporin Nup155; nucleoporin Nup160; Nucleoporin Nup205; nucleoporin Nup210; Nucleoporin Nup214; nucleoporin Nup35; Nucleoporin Nup50; nucleoporin NUP53; Nucleoporin Nup62; nucleoporin Nup75; Nucleoporin Nup84; nucleoporin NUP85; Nucleoporin Nup88; nucleoporin Nup93; Nucleoporin Nup96; Nucleoporin Nup98; nucleoprotein 50; Nucleoprotein 50kD; NUCZINK; Nup107; NUP120; Nup133; Nup153; NUP155; Nup160; NUP196; Nup205; NUP210; Nup214; Nup35; Nup50; NUP53; NUP62; NUP75; Nup84; NUP85; NUP88; Nup93; Nup96; Nup98; NUP98/PHF23 fusion 2 protein; Nup98-Nup96; Nupc1; p105; P140; p62; Pcnt1; pericentrin 1; pericentrin-1; POM210; pore membrane protein of 210 kDa; Prei2; preimplantation protein 2; PRO1146; Protein CAN; PSEC0245; RGD1304977; Rtp60; SNDI
Gene Symbols NUP107, NUP133, NUP153, NUP155, NUP160, NUP205, NUP210, NUP214, NUP35, NUP50, NUP62, NUP85, NUP88, NUP93, NUP98
Host Species Mouse
Immunogen Yeast nuclear preparations.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103468, 10762, 113929, 116555, 117021, 129401, 170762, 18141, 18226, 19069, 218210, 227720, 23165, 23225, 23279, 234865, 23636, 25281, 25497, 269966, 287830, 291874, 292085, 295692, 296634, 311182, 362335, 445007, 4927, 4928, 54563, 55746, 57122, 58958, 59015, 65274, 69482, 70699, 71805, 79902, 8021, 81738, 9631, 9688, 9972
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.