Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTH1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$181.72 - $492.89
Specifications
| Antigen | NTH1 |
|---|---|
| Concentration | 1.1 mg/mL |
| Dilution | Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100108SS
![]() |
Novus Biologicals
NB100108SS |
0.05 mL |
Each for $181.72
|
|
|||||
NB100108
![]() |
Novus Biologicals
NB100108 |
0.2 mL |
Each for $492.89
|
|
|||||
Description
NTH1 Polyclonal specifically detects NTH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NTH1 | |
| Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
| NTHL1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse and rat. Immunogen sequence has 93% identity with bovine proteins. |
| 1.1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P78549 | |
| 4913 | |
| A peptide derived from human NTH1, conjugated to KLH. [UniProt# P78549] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 37 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title