Learn More
Aviva Systems Biology Corp NR2F2 Antibody - C-terminal region (ARP39466_T100) | 100uL
Supplier: Aviva Systems Biology Corp ARP39466_T100
This is a rabbit polyclonal antibody against NR2F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR2F2. Alternate names: ARP1, NF-E3, NR2F1, SVP40, COUPTFB, TFCOUP2, COUPTFIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.