Learn More
Invitrogen™ ELISA TMB Stabilized Chromogen
For use with all sandwich ELISAs which utilize Horseradish Peroxidase (HRP) as detecting enzyme
$100.00 - $642.00
Description
- Formulation: Liquid
- Available in two sizes
- Detection method: Colormetric
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping Condition: Wet Ice
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.