Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (20X)

Catalog No. NP0001
Encompass_Preferred
Click to view available options
Quantity:
500 mL
5 L

Catalog No. NP0001

Supplier: Invitrogen™ NP0001

Only null left
Add to Cart
Edge
Add to Cart

Includes

NuPAGE MOPS SDS Running Buffer (20X)

Requires

The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.

Made with high-purity reagents and are strictly quality controlled.

NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE™ MOPS SDS Running Buffer is recommended for separating small- to medium-sized proteins.

  • Get consistent, convenient, high-quality results
  • Pre-mixed NuPAGE buffers are convenient way to ensure high-quality, consistent electrophoresis results

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 20X
pH 7.7
Physical Form Liquid
Product Line NuPAGE
Quantity 500 mL
Formulation 50 mM MOPS, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.7, 0.01-0.09% N,N-dimethylformamide
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.