Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene (Certified ACS), Fisher Chemical™

Catalog No. n91i500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass/Poison Pack, EcoSafPak™
Glass Bottle

Catalog No. N91I-500

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 5°C
Color Yellow
Density 1.205g/cm³
Boiling Point 210°C
Packaging Glass Bottle
Quantity 500 mL
Residue after Evaporation 0.005% max.
Vapor Pressure 0.2mbar at 20°C
Formula Weight 123.11
Percent Purity ≥99.0%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.