Learn More
Med Vet International Nipro Syringe & Needle, 3mL, Luer Lock, 20G X 1 in, Hypodermic, 100/BX, JD+03L2025
Supplier:  Med Vet International JD03L2025
Encompass_Preferred
Nipro JD+03L2025 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Nipro JD+03L2025 Syringe & Needle Combo comes with a 20G X 1 ' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. NC1545300
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.