Learn More
Lkt Laboratories CAS 98-92-0, ≥98%, Amide form of vitamin B3 required for production of NAD and NADP. It modulates GABA activity and is used to treat acne vulgaris and rosacea.
Supplier: Lkt Laboratories N3310
Nicotinamide is an amide form of vitamin B3 that is incorporated into NAD and NADP as oxidation-reduction coenzymes. Nicotinamide exhibits anti-inflammatory and anxiolytic activities; it is clinically used to treat acne vulgaris and rosacea. Commercially, nicotinamide is used in skin whitening treatments as it decreases skin pigmentation. Nicotinamide also binds GABA receptors, decreasing aggressive behavior.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.