Learn More
New England Biolabs, Inc. NEBNext Enzymatic Methyl-seq Conversion Module – 96 reactions
Supplier: New England Biolabs, Inc. E7125L
NEBNext Enzymatic Methyl-seq (EM-seq) is a new method for identification of 5-mC and 5-hmC. While bisulfite sequencing has been the gold standard for methylome analysis, this conversion treatment damages DNA, resulting in fragmentation, loss and bias. The NEBNext Enzymatic Methyl-seq Conversion Module provides an enzyme-based alternative to bisulfite conversion. For use in Illumina sequencing workflows, we recommend use of the NEBNext Enzymatic Methyl-seq Kit instead of this module. The full kit includes the EM-seq Adaptor, which has been optimized for high performance in the EM-seq workflow, as well as the high-efficiency Ultra II library prep reagents, and these components are not included in the Conversion Module.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.