Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NERL™ High Purity Water

Catalog No. 23249589
Click to view available options
Quantity:
1 gal.
5 gal.
This item is not returnable. View return policy
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation

Catalog No. 23-249-589

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.

Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.

Contains no preservatives

Ordering Information:

Certificate of analysis available for each case

TRUSTED_SUSTAINABILITY

Specifications

pH 5 to 7
Grade NERL
Quantity 1 gal.
ShelfLife 12 Months
Recommended Storage 15° to 30°C

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.