Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit v2 (Human/Mouse/Rat) – 96 reactions
SDP
Supplier:  New England Biolabs, Inc. E7400X
Encompass
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human, mouse and rat total RNA preparations. This product is suitable for both intact and degraded RNA (e.g. FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications.
Catalog No. 50-202-4023
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.