Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE

Catalog No. 02925113
Click to view available options
Thread Size (Metric):
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Product Type:
Replacement Cap
Spout Cap
Color:
Amber
Natural
White

Catalog No. 02-925-113

Supplier: Thermo Scientific™ 7121510200

Only null left
Add to Cart
Add to Cart

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Bottle Replacement Closures
Material HDPE
No. per Case 12
Thread Size (Metric) 20 mm
Mouth Narrow
Shape Round
Product Type Replacement Cap
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.