Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles
GSA_VAGREENER_CHOICE

Catalog No. 02923g
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Closure Size:
38-430 mm
53B

Catalog No. 02-923G

Supplier: Thermo Scientific™ 22020005

Only null left
Add to Cart
Add to Cart

Collect and store distilled water in bottles that offer flexibility, impact resistance and visibility of contents.

Store chemicals and standards and collect and store distilled water. Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low-density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contiminate your reagents

Includes:

Leakproof† polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 gal., 2 L
Capacity (Metric) 2 L
Capacity (English) 0.5 gal.
Color Translucent
Description Large Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 25 mm
Material LDPE
No. per Case 6
No. per Pack 1
Sterility Non-sterile
Closure Color White
Neck Finish 38 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 249 mm
Product Type Narrow Mouth Bottle
Height (English) 9.8 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 121 mm
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.