Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Plastic Graduated Cylinder Variety Pack
GREENER_CHOICE

Catalog No. 02540270
Click to view available options
Capacity (Metric):
10, 25, 50, 100, 250, 500, 1000 mL

Catalog No. 02-540-270

Supplier: Thermo Scientific™ 36621234

Only null left
Add to Cart
Add to Cart

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Seven sizes, from 10 to 1000mL.

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Thermo Scientific™ Nalgene™ Plastic Graduated Cylinder Variety Packs include one each 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL cylinders. All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. No meniscus allows accurate readings every time. The large base keeps cylinders stable on the benchtop preventing tipping hazards—won't break or chip like glass labware. Suitable for food and beverage use.

  • Includes seven cylinders, one each size: 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL
  • Guaranteed to meet requirements of ASTM Class B, E1272 for accuracy
  • No meniscus to confuse readings—eliminates guesswork
  • Large blue octagonal base prevents rolling and tipping
  • Generous pour spout provides ease in dispensing liquids
  • Molded-in graduations are silk-screened for easy reading
  • Excellent chemical resistance for a wide variety of solutions
  • Impact resistant at temperatures above -40°C
  • Autoclavable/Graduated
Compliance:
  • Meets ASTM Class B, E1272 Class B and 21 CFR Part 177.1520 for food/beverage use
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10, 25, 50, 100, 250, 500, 1000 mL
Material PPCO
Item Description Nalgene Grad Cylinder Variety Pack, PPCO
Certifications/Compliance ASTM, CFR 21 Part 177.1520, E1272
No. per Pack 7
Scale mL
Includes Blue Polypropylene Base
Autoclavable Autoclavable
Max. Temperature 121°C
Subdivision Yes
Shape Round
Max. Temperature (English) 250°F
For Use With (Application) Food and Beverage Use, Refreshing an Existing Lab
Base Type Octagonal
Type Graduated Cylinder Set
Show More Show Less

Notes:

  • Autoclaving will affect graduation accuracy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.