Learn More
Sigma Aldrich Fine Chemicals Biosciences N,N,N,N-Tetramethylethylenediamine, 110-18-9, MFCD00008335, 25mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T928125ML
Encompass
Linear Formula (CH3)2NCH2CH2N(CH3)2, ~99%, Molecular Weight 116.20, BioReagent, suitable for electrophoresis, Synonym: 1,2-Bis(dimethylamino)ethane, TEMED, TMEDA
Catalog No. 50-165-6850
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.