Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2,6-diisopropylphenyl)-1,6,7,12-tetraphenoxy-3,4,9,10-perylenetetracarboxylic Diimide 95.0+%, TCI America™
SDP

Catalog No. B43941G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. B43941G

Supplier: TCI America B43941G

Chemical Identifiers

CAS 123174-58-3
Molecular Formula C72H58N2O8
Molecular Weight (g/mol) 1079.262
InChI Key ZZSIDSMUTXFKNS-UHFFFAOYSA-N
PubChem CID 16172388
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C(=O)C3=CC(=C4C5=C(C=C6C7=C5C(=C(C=C7C(=O)N(C6=O)C8=C(C=CC=C8C(C)C)C(C)C)OC9=CC=CC=C9)C1=C(C=C(C3=C41)C2=O)OC1=CC=CC=C1)OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Red
Quantity 1 g
Formula Weight 1079.26
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2,6-diisopropylphenyl)-1,6,7,12-tetraphenoxy-3,4,9,10-perylenetetracarboxylic Diimide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.