Learn More
N-Benzylmethylamine, 97%
Description
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
Specifications
Specifications
| Melting Point | -55°C |
| Density | 0.94 |
| pH | 10 |
| Boiling Point | 184°C to 188°C |
| Flash Point | 77°C (170°F) |
| Odor | Amine-like |
| Refractive Index | 1.523 |
| Linear Formula | C6H5CH2NHCH3 |
| Quantity | 100 mL |
| UN Number | UN2922 |
| Show More |
Safety and Handling
| Hazard Category | H227-H301-H314-H335 |
| Hazard Statement | GHS H Statement H301-H314-H318-H227 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 203-133-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.