Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. H126225G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Catalog No. H126225G

Supplier: TCI America H126225G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7646-67-5
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
InChI Key UUORTJUPDJJXST-UHFFFAOYSA-N
PubChem CID 193628
IUPAC Name N-(2-hydroxyethyl)prop-2-enamide
SMILES C=CC(=O)NCCO

Specifications

Boiling Point 130°C
Quantity 25 g
Formula Weight 115.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ)
EINECSNumber (2)-1023
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.