Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYST2 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00011143B01P
Description
Mouse polyclonal antibody raised against a full-length human MYST2 protein.
- High specificity
- High reactivity
Specifications
| MYST2 purified MaxPab mouse | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MYST2 protein. | |
| KAT7 | |
| HBO1/HBOA/KAT7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11143 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007067.3 | |
| MYST2 | |
| MYST2 (NP_008998.1, 1 a.a. ∼ 611 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction