Description
- Desiccation is prevented with a built in humidity chamber and the culture can be incubated for long periods of time
- Device fits conveniently on the microscope stage for easy observation without disturbing the fungal structures
- Potato Flake Agar
- Includes cover slip and humidifier
Specifications
Specifications
| Media Type | Potato Flake Agar |
| Target Organism Class | Fungi |
| Includes | 1 Agar Block, 1 Humidifying Chamber, 1 Plastic Coverslip, 1 Plastic Lid |
| No. per Pack | 5 Trays |
| Packaging Type | Box |