Learn More
Research Products International Corp MultiFlex Micro-Capillary Pipet Tips for Gel Loading, Non-Sterile, RNase and DNase Free, 0.5-200ul, Round Tip, Bulk Pack 1000 per Package
Supplier: Research Products International Corp 148010
MultiFlex tips are designed for precise loading of gels run in horizontal and vertical electrophoresis equipment. Other applications include DNA sequencing, nick translations, restriction digests and radioactive labelling. Revolutionary design provides a tight positive seal on small volume (1-200µl) sizes of air displacement micro-pipettors. MultiFlex tips fit all popular micro-pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA and Volac.
Tips are constructed of nucleated polypropylene and can be autoclaved to 120°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.