Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Respiratory Protection Instruments and Accessories

Catalog No. p-4014615 Shop All MSA Products
Change view
Click to view available options
Type:
Adhesive Backing Strip
Isobutylene Cylinder
For Use With (Equipment):
Respiratory Protection Instruments
Suspender clip with msa solaris multigas detector respiratory protection instruments
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
19-716-781 Isobutylene Cylinder Respiratory Protection Instruments
19-716-776 Adhesive Backing Strip Suspender clip with msa solaris multigas detector respiratory protection instruments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-716-781 Supplier MSA™ Supplier No. 10048279
Only null left
Add to Cart
Add to Cart

Unique clip designs keep the unit attached during even the roughest use

MSA™ Econo-Cal Isobutylene Cylinder:

  • 34L capacity
  • 100ppm
Solaris Backstrap Adhesive Clip:
  • For use with MSA Solaris multigas detector

Specifications

For Use With (Equipment) Respiratory Protection Instruments
Type Isobutylene Cylinder
Pressure Range 500 psi
Quantity 1 Ea.
Weight (English) 11 oz.
Weight (Metric) 0.31 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.