Learn More
MSA™ CABA Pressure Demand Regulator Accessory, Diaphragm
Catalog No. 189993454 Shop All MSA Products
Click to view available options
:
Diaphragm; Pressure demand regulator
Catalog No. 18-999-3454
Supplier: MSA™ 462282
Only null left
Add to Cart
Add to Cart
Compressed airline breathing apparatus (CABA) corresponding accessory provides a supply of safe air wherever air is irrespirable or oxygen deficient.
  • Accessory for compressed airline breathing apparatus (CABA)
  • Allows for independent breathing in irrespirable conditions, including contaminated ambient air and oxygen deficiency
  • Suitable for long-duration work and special applications
  • Modular design for multiple configurations
  • ASV (automatic switch valve) supplies air even if the airline is disconnected or the external air source fails
Specifications
Certifications/Compliance EN139
For Use With (Equipment) Compressed Airline Breathing Apparatus
Model 462282
Product Type Diaphragm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.