Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LSM™ Lymphocyte Separation Medium

Catalog No. icn50494 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 x 100 mL

Catalog No. ICN50494

Supplier: MP Biomedicals™ 0850494

Only null left
Add to Cart
Add to Cart

LSM™ Lymphocyte Separation Medium is a high performance density gradient medium for monocyte separation.

  • LSM is a sterile-filtered solution of a sucrose polymer and diatrizoate salts
  • Formulated for isolation of mononuclear cells from defibrinated or heparinized whole human blood
  • One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
  • Maximum yield of mononuclear cells
  • > 96% cell viability of lymphocytes
  • Easy one-step centrifugation
  • Low endotoxins
  • Autoclave sterilization
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Cell Culture Medium
Sterility Sterile
Quantity 5 x 100 mL
pH 7.23
Content And Storage 15° to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.