Learn More
Newcomer Supply Movat-Russell Modified Pentachrome Stain Kit, 250 ml kit
Supplier: Newcomer Supply 9150A
Newcomer Supply Movat-Russell Modified Pentachrome Stain Kit provides a single staining procedure that demonstrates five connective tissue elements; mucin, fibrin, elastic fibers, muscle, and collagen.