Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-MAP2K6, Clone: 3H12C8, Abnova™

Catalog No. 89108994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
89-108-994 100 μL
1 options

Catalog No. 89-108-994

Supplier: Abnova Corporation MAB10794

Only null left
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone 3H12C8
Description Mouse monoclonal antibody raised against partial recombinant MAP2K6.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene Alias MAPKK6/MEK6/MKK6/PRKMK6/SAPKK3
Gene Symbols MAP2K6
Immunogen Recombinant protein corresponding to human MAP2K6.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5608
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.