Learn More
Invitrogen™ Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 647 Conjugate
Description
Lectin GS-II is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or Β-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The far red-fluorescent lectin GS-II Alexa Fluor 647 conjugate can be used in fluorescence microscopy or flow cytometry.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging, Microbial Tracking
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 500 μg |
| Source | Africa Shrub legume |
| Conjugate | Alexa Fluor 647 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Lectins |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.